accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 139742-50-0

NNOHO

CAS RN 139742-50-0

IUPAC Name: 4-[(1H-1,3-benzodiazol-1-yl)methyl]benzoic acid
Synonyms: 4-(1,3-benzodiazol-1-ylmethyl)benzoic acid
Molecular Weight (g/mol): 252.27
Molecular Formula: C15H12N2O2
InChi Key: KLIANJXCGMBICV-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(CN2C=NC3=CC=CC=C23)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results