accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 139768-71-1

IUPAC Name: 1-(benzyloxy)-4-isothiocyanatobenzene
Synonyms: 1-(benzyloxy)-4-isothiocyanatobenzene
Molecular Weight (g/mol): 241.31
Molecular Formula: C14H11NOS
InChi Key: OQXRBXAFSXVCCO-UHFFFAOYSA-N
SMILES: S=C=NC1=CC=C(OCC2=CC=CC=C2)C=C1
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results