accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

(S)(S)HNOHOOHCH3OHO∗O(S)(S)HNOHO∗CH3OHO∗O(S)(S)HNOHO∗CH3OHOOHn

CAS RN 1398-61-4

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): —
Molecular Formula: (C8H13NO5)nC16H28N2O11
InChi Key: —
SMILES: CC(=O)N[C@@H]1C(O)OC(CO)C(-*-O-*-C2OC(CO)C(-*-O-*-C3OC(CO)C(O)[C@@H](O)[C@@H]3NC(C)=O)[C@@H](O)[C@@H]2NC(C)=O)[C@H]1O
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)