CAS RN 13986-18-0
IUPAC Name:
zinc(2+) tetrahydrate difluoride
Synonyms:
zinc(2+) tetrahydrate difluoride
Molecular Weight (g/mol):
175.44
Molecular Formula:
F2H8O4Zn
InChi Key:
OEUUYPRXVRBIFO-UHFFFAOYSA-L
SMILES:
O.O.O.O.[F-].[F-].[Zn++]
1
–
1
of
1
results
1
Zinc fluoride tetrahydrate, 98%, Thermo Scientific Chemicals
CAS: 13986-18-0 Molecular Formula: F2H8O4Zn Molecular Weight (g/mol): 175.437 MDL Number: MFCD00149892 InChI Key: OEUUYPRXVRBIFO-UHFFFAOYSA-L Synonym: zinc fluoride tetrahydrate,zinc fluoride, tetrahydrate,unii-sq7815wxfb,zinc difluoride tetrahydrate,sq7815wxfb,zinc fluoride tetrahydrate mi,acmc-1c1qo,ksc497c6d,zinc, ion zn2+ tetrahydrate difluoride,zinc fluoride, tetrahydrate, PubChem CID: 44717730 IUPAC Name: zinc;difluoride;tetrahydrate SMILES: O.O.O.O.[F-].[F-].[Zn+2]