accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OSOOH3C

CAS RN 139926-22-0

IUPAC Name: methyl 3-ethoxythiophene-2-carboxylate
Synonyms: methyl 3-ethoxythiophene-2-carboxylate
Molecular Weight (g/mol): 186.23
Molecular Formula: C8H10O3S
InChi Key: JAGLXKOXARHKBR-UHFFFAOYSA-N
SMILES: CCOC1=C(SC=C1)C(=O)OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results