accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3BrSOOCl

CAS RN 139937-37-4

IUPAC Name: 4-bromo-2-methylbenzene-1-sulfonyl chloride
Synonyms: 4-bromo-2-methylbenzenesulfonyl chloride
Molecular Weight (g/mol): 269.54
Molecular Formula: C7H6BrClO2S
InChi Key: KNBSFUSBZNMAKU-UHFFFAOYSA-N
SMILES: CC1=C(C=CC(Br)=C1)S(Cl)(=O)=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)