accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 140-31-8

IUPAC Name: 2-(piperazin-1-yl)ethan-1-amine
Synonyms: aminoethylpiperazine
Molecular Weight (g/mol): 129.21
Molecular Formula: C6H15N3
InChi Key: IMUDHTPIFIBORV-UHFFFAOYSA-N
SMILES: NCCN1CCNCC1
Molecular Weight (g/mol) 
  • (2)
  • (5)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (5)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)