accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H2NF

CAS RN 140-75-0

IUPAC Name: 1-(4-fluorophenyl)methanamine
Synonyms: 4-fluorobenzylamine
Molecular Weight (g/mol): 125.15
Molecular Formula: C7H8FN
InChi Key: IIFVWLUQBAIPMJ-UHFFFAOYSA-N
SMILES: NCC1=CC=C(F)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)