CAS RN 140-95-4
IUPAC Name:
1,3-bis(hydroxymethyl)urea
Synonyms:
dimethylol urea
Molecular Weight (g/mol):
120.11
Molecular Formula:
C3H8N2O3
InChi Key:
QUBQYFYWUJJAAK-UHFFFAOYSA-N
SMILES:
OCNC(=O)NCO
1
–
1
of
1
results
1
1,3-Bis(hydroxymethyl)urea, tech. 90%
CAS: 140-95-4 Molecular Formula: C3H8N2O3 Molecular Weight (g/mol): 120.11 MDL Number: MFCD00014414 InChI Key: QUBQYFYWUJJAAK-UHFFFAOYSA-N Synonym: 1,3-bis hydroxymethyl urea,dimethylolurea,oxymethurea,n,n'-dimethylolurea,methural,carbamol,caurite,metural,urofix,protesine dmu PubChem CID: 8827 IUPAC Name: 1,3-bis(hydroxymethyl)urea SMILES: OCNC(=O)NCO