accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1400-61-9

IUPAC Name: (1S,15S,16R,17R,18S,19E,21E,25E,27E,29E,31E)-33-{[(2S,3S,4S,5S,6R)-4-amino-3,5-dihydroxy-6-methyloxan-2-yl]oxy}-1,3,4,7,9,11,17,37-octahydroxy-15,16,18-trimethyl-13-oxo-14,39-dioxabicyclo[33.3.1]nonatriaconta-19,21,25,27,29,31-hexaene-36-carboxylic acid
Synonyms: nystatin
Molecular Weight (g/mol): 926.11
Molecular Formula: C47H75NO17
InChi Key: VQOXZBDYSJBXMA-RKEBNKJGSA-N
SMILES: C[C@H]1O[C@H](OC2CC3O[C@@](O)(CC(O)C3C(O)=O)CC(O)C(O)CCC(O)CC(O)CC(O)CC(=O)O[C@@H](C)[C@H](C)[C@H](O)[C@@H](C)\C=C\C=C\CC\C=C\C=C\C=C\C=C\2)[C@@H](O)[C@@H](N)[C@@H]1O
Quantity 
  • (2)
  • (3)
  • (2)
  • (3)
  • (4)
  • (1)
Molecular Weight (g/mol) 
  • (6)
  • (7)
Physical Form 
  • (2)
  • (2)
  • (1)
Type 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (5)
  • (2)
  • (3)
  • (2)
  • (3)
  • (4)
  • (1)
  • (6)
  • (7)
  • (2)
  • (2)
  • (1)
  • (4)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)