accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CSNH3CN

CAS RN 14001-63-9

IUPAC Name: 4-methyl-2-(methylsulfanyl)pyrimidine
Synonyms: 4-methyl-2-(methylsulfanyl)pyrimidine
Molecular Weight (g/mol): 140.2
Molecular Formula: C6H8N2S
InChi Key: UCERVHYBSTYCQS-UHFFFAOYSA-N
SMILES: CSC1=NC=CC(C)=N1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)