accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOCH3H3CHO

CAS RN 14002-80-3

IUPAC Name: methyl 3-hydroxy-2,2-dimethylpropanoate
Synonyms: methyl 3-hydroxy-2,2-dimethylpropanoate
Molecular Weight (g/mol): 132.16
Molecular Formula: C6H12O3
InChi Key: KJRFTNVYOAGTHK-UHFFFAOYSA-N
SMILES: COC(=O)C(C)(C)CO
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)