accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OH2N

CAS RN 14006-32-7

IUPAC Name: 1-(4-phenyloxan-4-yl)methanamine
Synonyms: 1-(4-phenyloxan-4-yl)methanamine
Molecular Weight (g/mol): 191.27
Molecular Formula: C12H17NO
InChi Key: IPYDNTFWMDEOBW-UHFFFAOYSA-N
SMILES: NCC1(CCOCC1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results