accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 14017-56-2

IUPAC Name: yttrium(3+) hexahydrate triperchlorate
Synonyms: yttrium(3+) hexahydrate triperchlorate
Molecular Weight (g/mol): 495.33
Molecular Formula: Cl3H12O18Y
InChi Key: IYECSZYJKQPOOA-UHFFFAOYSA-K
SMILES: O.O.O.O.O.O.[Y+3].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results