accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOOOOOHNONHOHNOHNHOOClOHNHOOOHOHHOOH2NHNOH3CCH3HNH3CHOClHOHOHOCH3NH2HOHCl(?)

CAS RN 1404-93-9

IUPAC Name: 48-({3-[(4-amino-5-hydroxy-4,6-dimethyloxan-2-yl)oxy]-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl}oxy)-22-(carbamoylmethyl)-5,15-dichloro-2,18,32,35,37-pentahydroxy-19-[4-methyl-2-(methylamino)pentanamido]-20,23,26,42,44-pentaoxo-7,13-dioxa-21,24,27,41,43-pentaazaoctacyclo[26.14.2.2³,⁶.2¹⁴,¹⁷.1⁸,¹².1²⁹,³³.0¹⁰,²⁵.0³⁴,³⁹]pentaconta-3,5,8(48),9,11,14,16,29(45),30,32,34,36,38,46,49-pentadecaene-40-carboxylic acid hydrochloride
Synonyms: 48-({3-[(4-amino-5-hydroxy-4,6-dimethyloxan-2-yl)oxy]-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl}oxy)-22-(carbamoylmethyl)-5,15-dichloro-2,18,32,35,37-pentahydroxy-19-[4-methyl-2-(methylamino)pentanamido]-20,23,26,42,44-pentaoxo-7,13-dioxa-21,24,27,41,43-pentaazaoctacyclo[26.14.2.2³,⁶.2¹⁴,¹⁷.1⁸,¹².1²⁹,³³.0¹⁰,²⁵.0³⁴,³⁹]pentaconta-3,5,8(48),9,11,14,16,29(45),30,32,34,36,38,46,49-pentadecaene-40-carboxylic acid hydrochloride
Molecular Weight (g/mol): 1485.72
Molecular Formula: C66H76Cl3N9O24
InChi Key: LCTORFDMHNKUSG-UHFFFAOYNA-N
SMILES: Cl.CNC(CC(C)C)C(=O)NC1C(O)C2=CC(Cl)=C(OC3=CC4=CC(OC5=C(Cl)C=C(C=C5)C(O)C5NC(=O)C(NC(=O)C4NC(=O)C(CC(N)=O)NC1=O)C1=CC(=C(O)C=C1)C1=C(O)C=C(O)C=C1C(NC5=O)C(O)=O)=C3OC1OC(CO)C(O)C(O)C1OC1CC(C)(N)C(O)C(C)O1)C=C2
Quantity 
  • (4)
  • (1)
  • (3)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (1)
  • (5)
Physical Form 
  • (1)
  • (4)
  • (1)
  • (1)
Color 
  • (1)
  • (2)
  • (3)
Type 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (4)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (5)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (5)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)