accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 14044-65-6

IUPAC Name: boron(3+) oxolane trihydride
Synonyms: borane-tetrahydrofuran
Molecular Weight (g/mol): 85.94
Molecular Formula: C4H11BO
InChi Key: NOBZAJKNWRXAKG-UHFFFAOYSA-N
SMILES: [H-].[H-].[H-].[B+3].C1CCOC1
Quantity 
  • (3)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results