accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HClCH3NH3COHOO(Z)

CAS RN 140462-76-6

IUPAC Name: hydrogen 2-[(2Z)-2-[3-(dimethylamino)propylidene]-9-oxatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-5-yl]acetic acid chloride
Synonyms: hydrogen olopatadine chloride
Molecular Weight (g/mol): 373.88
Molecular Formula: C21H24ClNO3
InChi Key: HVRLZEKDTUEKQH-NOILCQHBSA-N
SMILES: [H+].[Cl-].CN(C)CC\C=C1\C2=CC=CC=C2COC2=CC=C(CC(O)=O)C=C12
Molecular Weight (g/mol) 
  • (5)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)