accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2SOOFF

CAS RN 140480-89-3

IUPAC Name: 3,5-difluorobenzene-1-sulfonamide
Synonyms: 3,5-difluorobenzenesulfonamide
Molecular Weight (g/mol): 193.17
Molecular Formula: C6H5F2NO2S
InChi Key: MKQPOVUFDWKPNO-UHFFFAOYSA-N
SMILES: NS(=O)(=O)C1=CC(F)=CC(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results