CAS RN 1405-97-6
CAS RN 1405-97-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Gramicidin (Technical Grade), TRC
CAS: 1405-97-6 Molecular Formula: C99H140N20O17 Molecular Weight (g/mol): 1882.3 Synonym: Gramoderm; SMILES: CCOc1c(C(=O)NCC2CN(CCO2)Cc3ccc(F)cc3)cc(Cl)c(c1)NC(=O)C(CC(O)=O)(O)CC(O)=O