CAS RN 14062-18-1
CAS RN 14062-18-1
IUPAC Name:
ethyl 2-(4-methoxyphenyl)acetate
Synonyms:
ethyl 2-(4-methoxyphenyl)acetate
Molecular Weight (g/mol):
194.23
Molecular Formula:
C11H14O3
InChi Key:
DOCCDOCIYYDLGJ-UHFFFAOYSA-N
SMILES:
CCOC(=O)CC1=CC=C(OC)C=C1
1
–
1
of
1
results
1
Ethyl 4-methoxyphenylacetate, 97%
CAS: 14062-18-1 Molecular Formula: C11H14O3 Molecular Weight (g/mol): 194.23 MDL Number: MFCD00040760 InChI Key: DOCCDOCIYYDLGJ-UHFFFAOYSA-N Synonym: ethyl 2-4-methoxyphenyl acetate,ethyl 4-methoxyphenylacetate,4-methoxyphenylacetic acid ethyl ester,benzeneacetic acid, 4-methoxy-, ethyl ester,ethyl p-methoxyphenylacetate,ethyl 4-methoxyphenyl acetate,4-methoxy-phenyl-acetic acid ethyl ester,acmc-1bnx2,homoanisic acid ethyl ester PubChem CID: 84174 IUPAC Name: ethyl 2-(4-methoxyphenyl)acetate SMILES: CCOC(=O)CC1=CC=C(C=C1)OC