accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2HNOH2N

CAS RN 14062-34-1

IUPAC Name: 3-aminobenzohydrazide
Synonyms: 3-aminobenzoylhydrazine
Molecular Weight (g/mol): 151.17
Molecular Formula: C7H9N3O
InChi Key: JSIVTKBYJLGBPY-UHFFFAOYSA-N
SMILES: NNC(=O)C1=CC=CC(N)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results