accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 14066-05-8

OClOOOOClOOOOClOOOSc

CAS RN 14066-05-8

IUPAC Name: scandium(3+) triperchlorate
Synonyms: scandium(3+) triperchlorate
Molecular Weight (g/mol): 343.29
Molecular Formula: Cl3O12Sc
InChi Key: UBVKRMCXDCYZQK-UHFFFAOYSA-K
SMILES: [Sc+3].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results