accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H3CCH3OONOH

CAS RN 140695-84-7

IUPAC Name: tert-butyl 3-(hydroxymethyl)piperidine-1-carboxylate
Synonyms: tert-butyl 3-(hydroxymethyl)piperidine-1-carboxylate
Molecular Weight (g/mol): 215.29
Molecular Formula: C11H21NO3
InChi Key: OJCLHERKFHHUTB-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)N1CCCC(CO)C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)