accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH3(S)(R)CH3O

CAS RN 14073-97-3

IUPAC Name: (2S,5R)-5-methyl-2-(propan-2-yl)cyclohexan-1-one
Synonyms: (-)-menthone (2S,5R)-menthone l-menthone
Molecular Weight (g/mol): 154.25
Molecular Formula: C10H18O
InChi Key: NFLGAXVYCFJBMK-BDAKNGLRSA-N
SMILES: CC(C)[C@@H]1CC[C@@H](C)CC1=O
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)
  • (2)
  • (4)
  • (6)