CAS RN 14080-23-0
IUPAC Name:
pyrimidine-2-carbonitrile
Synonyms:
2-pyrimidinecarbonitrile
Molecular Weight (g/mol):
105.1
Molecular Formula:
C5H3N3
InChi Key:
IIHQNAXFIODVDU-UHFFFAOYSA-N
SMILES:
N#CC1=NC=CC=N1
1
–
1
of
1
results
1
2-Pyrimidinecarbonitrile, 98%
CAS: 14080-23-0 Molecular Formula: C5H3N3 Molecular Weight (g/mol): 105.10 MDL Number: MFCD00160513 InChI Key: IIHQNAXFIODVDU-UHFFFAOYSA-N Synonym: 2-cyanopyrimidine,2-pyrimidinecarbonitrile,2-cyano-pyrimidine,2-cyano pyrimidine,cyanopyrimidine,provastatin,pyrimidinecarbonitrile,zlchem 283,pubchem7042,pyrimidin-2-carbonitrile PubChem CID: 2757979 IUPAC Name: pyrimidine-2-carbonitrile SMILES: N#CC1=NC=CC=N1