accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BOOH3CCH3H3CH3CNNCH3F

CAS RN 1408088-34-5

IUPAC Name: 1-[2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-4-methylpiperazine
Synonyms: 1-[2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-4-methylpiperazine
Molecular Weight (g/mol): 320.22
Molecular Formula: C17H26BFN2O2
InChi Key: RFECQDXNBWGCRR-UHFFFAOYSA-N
SMILES: CN1CCN(CC1)C1=C(F)C=C(C=C1)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results