accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

COCOCONOCo

CAS RN 14096-82-3

IUPAC Name: tris(methanidylidyneoxidanium) oxoazanide cobalt
Synonyms: tris(carbon monoxide) hyponitrite cobalt
Molecular Weight (g/mol): 172.97
Molecular Formula: C3CoNO4
InChi Key: CVCSGXJPONFHRC-UHFFFAOYSA-N
SMILES: [Co].[N-]=O.[C-]#[O+].[C-]#[O+].[C-]#[O+]
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)