accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 141-01-5

FeOOOO(Z)

CAS RN 141-01-5

IUPAC Name: λ²-iron(2+) (2Z)-but-2-enedioate
Synonyms: ferrous fumarate
Molecular Weight (g/mol): 169.9
Molecular Formula: C4H2FeO4
InChi Key: PMVSDNDAUGGCCE-ODZAUARKSA-L
SMILES: [Fe++].[O-]C(=O)\C=C/C([O-])=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results