accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 141-28-6

H3COOOOCH3

CAS RN 141-28-6

IUPAC Name: 1,6-diethyl hexanedioate
Synonyms: diethyl adipate diethylester kyseliny adipove diethylhexanedioate
Molecular Weight (g/mol): 202.25
Molecular Formula: C10H18O4
InChi Key: VIZORQUEIQEFRT-UHFFFAOYSA-N
SMILES: CCOC(=O)CCCCC(=O)OCC
Quantity 
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)