accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 141-32-2

IUPAC Name: butyl prop-2-enoate
Synonyms: butyl acrylate butyl-(2,3-14c)-acrylate butylester kyseliny akrylove butyl-(2,3-14C)-acrylate butyl acrylate (substance) butyl acrylate (product)
Molecular Weight (g/mol): 128.17
Molecular Formula: C7H12O2
InChi Key: CQEYYJKEWSMYFG-UHFFFAOYSA-N
SMILES: CCCCOC(=O)C=C
Molecular Weight (g/mol) 
  • (4)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)