accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 141-90-2

IUPAC Name: 2-sulfanylidene-1,2,3,4-tetrahydropyrimidin-4-one
Synonyms: thiouracil 2-mercapto-pyrimidin-4-ol
Molecular Weight (g/mol): 128.15
Molecular Formula: C4H4N2OS
InChi Key: ZEMGGZBWXRYJHK-UHFFFAOYSA-N
SMILES: O=C1NC(=S)NC=C1
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)
Certifications/Compliance 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)