accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 141-94-6

H3CCH3NNCH3CH3CH3H2N

CAS RN 141-94-6

IUPAC Name: 1,3-bis(2-ethylhexyl)-5-methyl-1,3-diazinan-5-amine
Synonyms: oraldene hexetidine rentschler brand drossadin hexetidine (product) triscol duranil aerosol hexoral caswell no. 033BB 海克替啶 Steri/Sol
Molecular Weight (g/mol): 339.61
Molecular Formula: C21H45N3
InChi Key: DTOUUUZOYKYHEP-UHFFFAOYNA-N
SMILES: CCCCC(CC)CN1CN(CC(CC)CCCC)CC(C)(N)C1
Molecular Weight (g/mol) 
  • (4)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)