CAS RN 14126-37-5
IUPAC Name:
dibromonickel; bis(triphenylphosphane)
Synonyms:
dibromonickel; bis(triphenylphosphine)
Molecular Weight (g/mol):
743.09
Molecular Formula:
C36H30Br2NiP2
InChi Key:
QEKXARSPUFVXIX-UHFFFAOYSA-L
SMILES:
Br[Ni]Br.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
Dibromobis(triphenylphosphine)nickel(II), 99%, Thermo Scientific Chemicals
CAS: 14126-37-5 Molecular Formula: C36H30Br2NiP2 Molecular Weight (g/mol): 743.09 MDL Number: MFCD00015865 InChI Key: QEKXARSPUFVXIX-UHFFFAOYSA-L Synonym: nickel ii bromide bis triphenylphosphine,nibr2 pph3 2,dibromobis triphenylphosphine nickel ii,bis triphenylphosphine nickel ii bromide,dibromobis triphenylphosphine nickel,dibromonickel; bis triphenylphosphine,bis triphenylphosphine dibromonickel ii PubChem CID: 11050914 IUPAC Name: dibromonickel;triphenylphosphane SMILES: Br[Ni]Br.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1