CAS RN 14167-18-1
CAS RN 14167-18-1
IUPAC Name:
λ²-cobalt(2+) 2-{[(2-{[(2-oxidophenyl)methylidene]amino}ethyl)imino]methyl}benzen-1-olate
Synonyms:
salcomine
Molecular Weight (g/mol):
325.23
Molecular Formula:
C16H14CoN2O2
InChi Key:
NPAQSKHBTMUERN-UHFFFAOYSA-L
SMILES:
[Co++].[O-]C1=CC=CC=C1C=NCCN=CC1=CC=CC=C1[O-]
1
–
1
of
1
results
1
N,N'-Bis(salicylidene)ethylenediaminecobalt(II), 96%
CAS: 14167-18-1 Molecular Formula: C16H14CoN2O2 Molecular Weight (g/mol): 325.23 MDL Number: MFCD00000009 InChI Key: NPAQSKHBTMUERN-UHFFFAOYSA-L Synonym: Salcomine PubChem CID: 57448789 SMILES: [Co++].[O-]C1=CC=CC=C1C=NCCN=CC1=CC=CC=C1[O-]