accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOOH

CAS RN 14199-15-6

IUPAC Name: methyl 2-(4-hydroxyphenyl)acetate
Synonyms: methyl 2-(4-hydroxyphenyl)acetate
Molecular Weight (g/mol): 166.18
Molecular Formula: C9H10O3
InChi Key: XGDZEDRBLVIUMX-UHFFFAOYSA-N
SMILES: COC(=O)CC1=CC=C(O)C=C1
Molecular Weight (g/mol) 
  • (5)
  • (3)
  • (2)
Quantity 
  • (3)
  • (3)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results