accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 142-04-1

IUPAC Name: hydrogen aniline chloride
Synonyms: hydrogen aniline chloride
Molecular Weight (g/mol): 129.59
Molecular Formula: C6H8ClN
InChi Key: MMCPOSDMTGQNKG-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NC1=CC=CC=C1
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)