accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH3OOOOCH3CH3(Z)

CAS RN 142-16-5

IUPAC Name: 1,4-bis(2-ethylhexyl) (2Z)-but-2-enedioate
Synonyms: di-2-ethylhexyl maleate diethylhexyl maleate
Molecular Weight (g/mol): 340.5
Molecular Formula: C20H36O4
InChi Key: ROPXFXOUUANXRR-YPKPFQOONA-N
SMILES: CCCCC(CC)COC(=O)\C=C/C(=O)OCC(CC)CCCC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results