accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 142-63-2

H2OH2OH2OH2OH2OH2ONHHN

CAS RN 142-63-2

IUPAC Name: piperazine hexahydrate
Synonyms: piperazine hexahydrate
Molecular Weight (g/mol): 194.23
Molecular Formula: C4H22N2O6
InChi Key: AVRVZRUEXIEGMP-UHFFFAOYSA-N
SMILES: O.O.O.O.O.O.C1CNCCN1
Molecular Weight (g/mol) 
  • (4)
  • (5)
Boiling Point 
  • (3)
  • (1)
  • (4)
Physical Form 
  • (4)
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (3)
  • (4)
  • (5)
  • (3)
  • (1)
  • (4)
  • (4)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (4)