accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1420-40-2

IUPAC Name: trimethyl[10-(trimethylazaniumyl)decyl]azanium diiodide
Synonyms: decamethonium iodide
Molecular Weight (g/mol): 512.3
Molecular Formula: C16H38I2N2
InChi Key: ARMLJSXXXFXSLQ-UHFFFAOYSA-L
SMILES: [I-].[I-].C[N+](C)(C)CCCCCCCCCC[N+](C)(C)C
Certifications/Compliance 
  • (1)
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results