accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 14205-39-1

IUPAC Name: methyl (2E)-3-aminobut-2-enoate
Synonyms: methyl (2E)-3-aminobut-2-enoate
Molecular Weight (g/mol): 115.13
Molecular Formula: C5H9NO2
InChi Key: XKORCTIIRYKLLG-ONEGZZNKSA-N
SMILES: COC(=O)\C=C(/C)N
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
Physical Form 
  • (3)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results