accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 14206-69-0

IUPAC Name: 3-amino-5-hydroxybenzoic acid hydrochloride
Synonyms: AHBA hydrochloride
Molecular Weight (g/mol): 189.6
Molecular Formula: C7H8ClNO3
InChi Key: CXESTILCPSBCGQ-UHFFFAOYSA-N
SMILES: Cl.NC1=CC(O)=CC(=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results