CAS RN 1421-89-2
IUPAC Name:
2-(dimethylamino)ethyl acetate
Synonyms:
2-(dimethylamino)ethyl acetate
Molecular Weight (g/mol):
131.18
Molecular Formula:
C6H13NO2
InChi Key:
GOLSFPMYASLXJC-UHFFFAOYSA-N
SMILES:
CN(C)CCOC(C)=O
1
–
1
of
1
results
1
2-Dimethylaminoethyl acetate, 98%, Thermo Scientific Chemicals
CAS: 1421-89-2 Molecular Formula: C6H13NO2 Molecular Weight (g/mol): 131.18 MDL Number: MFCD00041912 InChI Key: GOLSFPMYASLXJC-UHFFFAOYSA-N Synonym: 2-dimethylamino ethyl acetate,dimethylaminoethyl acetate,dimethylaminoethanol acetate,2-dimethylaminoethanol acetate,acetic acid, 2-dimethylamino ethyl ester,acetic acid 2-dimethylaminoethyl ester,2-dimethylamino ethanol acetic ester,acmc-1c2gl,2-dimethylaminoethyl=acetate,2-dimethylamino ethylacetate PubChem CID: 15013 IUPAC Name: 2-(dimethylamino)ethyl acetate SMILES: CN(C)CCOC(C)=O