accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

SN

CAS RN 1421261-80-4

IUPAC Name: 2-(naphthalen-2-yl)-4-(5,6,7,8-tetrahydronaphthalen-2-yl)-1,3-thiazole
Synonyms: 2-(naphthalen-2-yl)-4-(5,6,7,8-tetrahydronaphthalen-2-yl)-1,3-thiazole
Molecular Weight (g/mol): 341.47
Molecular Formula: C23H19NS
InChi Key: ISODJAPESPIWDI-UHFFFAOYSA-N
SMILES: C1CCC2=C(C1)C=CC(=C2)C1=CSC(=N1)C1=CC=C2C=CC=CC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results