accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H3CNOFF

CAS RN 1421262-76-1

IUPAC Name: N-benzyl-2,6-difluoro-N-(propan-2-yl)benzamide
Synonyms: N-benzyl-2,6-difluoro-N-isopropylbenzamide
Molecular Weight (g/mol): 289.33
Molecular Formula: C17H17F2NO
InChi Key: WMGHZTPHABWXOF-UHFFFAOYSA-N
SMILES: CC(C)N(CC1=CC=CC=C1)C(=O)C1=C(F)C=CC=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results