CAS RN 1421263-60-6
IUPAC Name:
N-benzyl-N-tert-butyl-4-chlorobutanamide
Synonyms:
N-benzyl-N-tert-butyl-4-chlorobutanamide
Molecular Weight (g/mol):
267.8
Molecular Formula:
C15H22ClNO
InChi Key:
UBCAUNFPEJQKIA-UHFFFAOYSA-N
SMILES:
CC(C)(C)N(CC1=CC=CC=C1)C(=O)CCCCl
1
–
1
of
1
results
1
N-Benzyl-N-tert-butyl-4-chlorobutyramide, 97%, Thermo Scientific™
CAS: 1421263-60-6 Molecular Formula: C15H22ClNO Molecular Weight (g/mol): 267.797 MDL Number: MFCD22683090 InChI Key: UBCAUNFPEJQKIA-UHFFFAOYSA-N Synonym: n-benzyl-n-tert-butyl-4-chlorobutyramide PubChem CID: 97030388 IUPAC Name: N-benzyl-N-tert-butyl-4-chlorobutanamide SMILES: CC(C)(C)N(CC1=CC=CC=C1)C(=O)CCCCl