accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 14215-68-0

IUPAC Name: N-[(3R,4R,5R,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide
Synonyms: GalNAc acetyl galactosamine N-acetyl-D-galactosamine D-GalNAc
Molecular Weight (g/mol): 221.21
Molecular Formula: C8H15NO6
InChi Key: OVRNDRQMDRJTHS-KEWYIRBNSA-N
SMILES: CC(=O)N[C@H]1C(O)O[C@H](CO)[C@H](O)[C@@H]1O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)