accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1422-54-4

CH3BrF

CAS RN 1422-54-4

IUPAC Name: 1-bromo-3-fluoro-2-methylbenzene
Synonyms: 1-bromo-3-fluoro-2-methylbenzene
Molecular Weight (g/mol): 189.03
Molecular Formula: C7H6BrF
InChi Key: DJGXPFQIMLEVPA-UHFFFAOYSA-N
SMILES: CC1=C(F)C=CC=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results