accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

PdClClNN

CAS RN 14220-64-5

IUPAC Name: palladium(2+) bis(benzonitrile) dichloride
Synonyms: palladium(2+) bis(benzonitrile) dichloride
Molecular Weight (g/mol): 383.57
Molecular Formula: C14H10Cl2N2Pd
InChi Key: WXNOJTUTEXAZLD-UHFFFAOYSA-L
SMILES: [Cl-].[Cl-].[Pd++].N#CC1=CC=CC=C1.N#CC1=CC=CC=C1
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)