accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 14221-01-3

IUPAC Name: tetrakis(triphenylphosphane) palladium
Synonyms: tetrakis(triphenylphosphine)palladium(0)
Molecular Weight (g/mol): 1155.59
Molecular Formula: C72H60P4Pd
InChi Key: NFHFRUOZVGFOOS-UHFFFAOYSA-N
SMILES: [Pd].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (9)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (4)
  • (2)
Physical Form 
  • (3)
  • (4)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (9)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (4)
  • (2)
  • (3)
  • (4)
  • (2)