accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOCH3OH3CNOO

CAS RN 14227-18-0

IUPAC Name: 1,3,5-trimethoxy-2-nitrobenzene
Synonyms: 1,3,5-trimethoxy-2-nitrobenzene
Molecular Weight (g/mol): 213.19
Molecular Formula: C9H11NO5
InChi Key: VWYAWLZEMLQGJH-UHFFFAOYSA-N
SMILES: COC1=CC(OC)=C(C(OC)=C1)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results